Orders LinkContactUsLinkComponent
Multi-Angle Light-Scattering Detector, LenS3™
Multi-Angle Light-Scattering Detector, LenS3™
Dodavatel:   TOSOH Bioscience
Multi-Angle Light-Scattering Detector, LenS3™
Dodavatel:  TOSOH Bioscience
Certifikáty
Multi-Angle Light-Scattering Detector, LenS3™
2 Dostupných variant níže
Product Details & Documents
Tosoh’s LenS3™ Multi-Angle Light-Scattering detector combines high-sensitivity MW and Rg determinations with flexibility and ease of use.
  • Characterisation of molecules down to 200 to 10⁷ Da and Rg down to <2 to >300 nm
  • Optics design for low noise and high scattering intensity
  • Compatible with GPC, SEC, HPLC and UHPLC systems
  • Compatible with all common solvents (THF, HFIP, aqueous salt buffers)
  • Modular software for simultaneous acquisition data analysis, sequence editing
  • Analysis modules for step-by-step guidance
  • Proprietary conical flow path
  • Laser: Diode, 20 mW ±5, wavelength 505 nm
  • Baseline noise: <1 mV (in THF at 1 ml/min)
  • Wetted materials: Teflon, PEEK, glass, stainless steel
The LenS3™ Multi-Angle Light-Scattering detector enables you to directly measure the molecular weight and radius of gyration of polymers, polysaccharides, and biopolymers such as proteins or delivery particles. Its high sensitivity allows the characterisation of low sample amounts, molecules with low MW or determination of Rg values downt to <10 nm. It serves you in combination with any SEC device such as GPC, HPLC or UHPLC instruments.
Maximum flow rate: 2 ml/min
Acquisition rate: 10 Hz
Dimensions (W×D×H): 36,5×48,5×13 mm
Weight: 16 kg
Product Family Options
Detector
Informace o produktu
  • Sekvence
    Dostupnost
    Price
  • TOYO0040000
    LenS3™ Multi-Angle Light-Scattering detector
    LenS3™ Multi-Angle Light-Scattering detector
    01
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 1 830 060,00 Kč
    1 830 060,00 Kč
    1 * 1 KS
       1 830 060,00 Kč
Conversion kit
Informace o produktu
  • Sekvence
    Dostupnost
    Price
  • TOYO0040001
    UHPLC conversion of LenS3™ MALS detector (only available with LenS3™)
    UHPLC conversion of LenS3™ MALS detector (only available with LenS3™)
    02
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 210 460,00 Kč
    210 460,00 Kč
    1 * 1 KS
       210 460,00 Kč
Services
Informace o produktu
  • Katalogové číslo
    Popis
    Sekvence
    Dostupnost
    Price
  • TOYO0046040
    Operational Qualification (OQ)
    03
    Přihlásit se pro dostupnost
    1 * 1 KS - 91 260,00 Kč
  • TOYO0046039
    Installation Qualification (IQ)
    02
    Přihlásit se pro dostupnost
    1 * 1 KS - 45 630,00 Kč
  • TOYO0046038
    Installation
    01
    Přihlásit se pro dostupnost
    1 * 1 KS - 91 260,00 Kč
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Stainless steel inline filter housing, Int.Ø: 13 mm
    Stainless steel inline filter housing, Int.Ø: 13 mm
    TOYO0040002
    08
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 560,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Post column filter 0,2 µm, 13 mm nylon
    Post column filter 0,2 µm, 13 mm nylon
    TOYO0040032
    02
    Přihlásit se pro dostupnost
    1 * 100 KS - 326,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Post pump filter 0,03 µm, 13 mm PES
    Post pump filter 0,03 µm, 13 mm PES
    TOYO0040048
    05
    Přihlásit se pro dostupnost
    1 * 100 KS - 3 770,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Post column filter 0,1 µm, 13 mm PTFE
    Post column filter 0,1 µm, 13 mm PTFE
    TOYO0040031
    01
    Přihlásit se pro dostupnost
    1 * 100 KS - 326,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Post column filter 0,2 µm, 13 mm PES
    Post column filter 0,2 µm, 13 mm PES
    TOYO0040049
    03
    Přihlásit se pro dostupnost
    1 * 100 KS - 3 770,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Post column filter 0,2 µm, 13 mm PTFE
    Post column filter 0,2 µm, 13 mm PTFE
    TOYO0040074
    04
    Přihlásit se pro dostupnost
    1 * 100 KS - 948,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Stainless steel frits, Int.Ø: 13 mm (for inline filter housing)
    Stainless steel frits, Int.Ø: 13 mm (for inline filter housing)
    TOYO0040030
    07
    Přihlásit se pro dostupnost
    1 * 10 KS - 1 520,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • UHPLC stainless steel inline micro filter
    UHPLC stainless steel inline micro filter
    TOYO0040043
    09
    Přihlásit se pro dostupnost
    1 * 5 KS - 7 560,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Post pump filter 0,1 µm, 13 mm nylon
    Post pump filter 0,1 µm, 13 mm nylon
    TOYO0040042
    06
    Přihlásit se pro dostupnost
    1 * 100 KS - 326,00 Kč