Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PE (Phycoerythrin))
Dodavatel: AAT Bioquest
Certifikáty
O tomto produktu
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Upozornení: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifikace
- Catalog No:
- AATB100331L0
- AATB100331L1
- AATB100331L2
- Symbol antigenu:
- CD3e
- CD3e
- CD3e
- Název antigenu:
- CD3e antigen
- CD3e antigen
- CD3e antigen
- Druh protilátky:
- Primární
- Primární
- Primární
- Klonovalita:
- Monoclonal
- Monoclonal
- Monoclonal
- Spojení:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Klon:
- SK7
- SK7
- SK7
- Reaktivita:
- Human
- Human
- Human
- Hostitel:
- Mouse
- Mouse
- Mouse
- ImunoFluorescence:
- Yes
- Yes
- Yes
- Průtoková cytometrie:
- Yes
- Yes
- Yes
- Čištění:
- Affinity chromatography
- Affinity chromatography
- Affinity chromatography
- Křížová adsorbce:
- No
- No
- No
- Skladovací teplota:
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- Transportní teplota:
- 20
- 20
- 20