Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 710)
Supplier: AAT Bioquest
Certificates
About this item
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: iFluor® 710
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB101910K1
- AATB101910K0
- Symbol antigenu:
- CD19
- CD19
- Název antigenu:
- CD19 antigen
- CD19 antigen
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Monoclonal
- Monoclonal
- Spojení:
- iFluor® 710
- iFluor® 710
- Klon:
- SJ25C1
- SJ25C1
- Reaktivita:
- Human
- Human
- Hostitel:
- Mouse
- Mouse
- ImunoFluorescence:
- Yes
- Yes
- Průtoková cytometrie:
- Yes
- Yes
- Čištění:
- Affinity chromatography
- Affinity chromatography
- Křížová adsorbce:
- No
- No
- Skladovací teplota:
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- Transportní teplota:
- 20
- 20