Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (PerCP)
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
: For research use only.
Type: Primary
Antigen: CD14 antigen
Clonality: Monoclonal
Clone: 61D3
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- AATB101411T0
- AATB101411T1
- AATB101411T2
- Symbol antigenu:
- CD14
- CD14
- CD14
- Název antigenu:
- CD14 antigen
- CD14 antigen
- CD14 antigen
- Druh protilátky:
- Primární
- Primární
- Primární
- Klonovalita:
- Monoclonal
- Monoclonal
- Monoclonal
- Spojení:
- PerCP (Peridinin-Chlorophyll Protein Complex)
- PerCP (Peridinin-Chlorophyll Protein Complex)
- PerCP (Peridinin-Chlorophyll Protein Complex)
- Klon:
- 61D3
- 61D3
- 61D3
- Reaktivita:
- Human
- Human
- Human
- Hostitel:
- Mouse
- Mouse
- Mouse
- ImunoFluorescence:
- Yes
- Yes
- Yes
- Průtoková cytometrie:
- Yes
- Yes
- Yes
- Čištění:
- Affinity chromatography
- Affinity chromatography
- Affinity chromatography
- Křížová adsorbce:
- No
- No
- No
- Skladovací teplota:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Transportní teplota:
- 20
- 20
- 20
Frequently Bought Together