Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (PE/iFluor® 700)
Supplier: AAT Bioquest
Certificates
About this item
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD5 antigen
Clonality: Monoclonal
Clone: L17F12
Conjugation: PE/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB100511X0
- AATB100511X1
- AATB100511X2
- Symbol antigenu:
- CD5
- CD5
- CD5
- Název antigenu:
- CD5 antigen
- CD5 antigen
- CD5 antigen
- Druh protilátky:
- Primární
- Primární
- Primární
- Klonovalita:
- Monoclonal
- Monoclonal
- Monoclonal
- Spojení:
- PE (Phycoerythrin)/iFluor® 700
- PE (Phycoerythrin)/iFluor® 700
- PE (Phycoerythrin)/iFluor® 700
- Klon:
- L17F12
- L17F12
- L17F12
- Reaktivita:
- Human
- Human
- Human
- Hostitel:
- Mouse
- Mouse
- Mouse
- ImunoFluorescence:
- Yes
- Yes
- Yes
- Průtoková cytometrie:
- Yes
- Yes
- Yes
- Čištění:
- Affinity chromatography
- Affinity chromatography
- Affinity chromatography
- Křížová adsorbce:
- No
- No
- No
- Skladovací teplota:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Transportní teplota:
- 20
- 20
- 20