Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (FITC)
Supplier: AAT Bioquest
Certificates
About this item
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Specifications
- Catalog No:
- AATB100411H1
- AATB100411H0
- Symbol antigenu:
- CD4
- CD4
- Název antigenu:
- CD4 antigen
- CD4 antigen
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Monoclonal
- Monoclonal
- Spojení:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Klon:
- RPA-T4
- RPA-T4
- Reaktivita:
- Human
- Human
- Hostitel:
- Mouse
- Mouse
- Izotyp:
- IgG1
- IgG1
- ImunoFluorescence:
- Yes
- Yes
- Průtoková cytometrie:
- Yes
- Yes
- Čištění:
- Affinity chromatography
- Affinity chromatography
- Křížová adsorbce:
- No
- No
- Skladovací teplota:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Transportní teplota:
- 20
- 20