Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 840)
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: iFluor® 840
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- AATB100520Q0
- AATB100520Q1
- Symbol antigenu:
- CD5
- CD5
- Název antigenu:
- CD5 antigen
- CD5 antigen
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Monoclonal
- Monoclonal
- Spojení:
- iFluor® 840
- iFluor® 840
- Klon:
- UCHT2
- UCHT2
- Reaktivita:
- Human
- Human
- Hostitel:
- Mouse
- Mouse
- ImunoFluorescence:
- Yes
- Yes
- Průtoková cytometrie:
- Yes
- Yes
- Čištění:
- Affinity chromatography
- Affinity chromatography
- Křížová adsorbce:
- No
- No
- Skladovací teplota:
- 2...8 °C with minimised light exposure, do not freeze
- 2...8 °C with minimised light exposure, do not freeze
- Transportní teplota:
- 20
- 20
Frequently Bought Together