Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 710)
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: iFluor® 710
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- AATB100330K0
- AATB100330K1
- Symbol antigenu:
- CD3e
- CD3e
- Název antigenu:
- CD3e antigen
- CD3e antigen
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Monoclonal
- Monoclonal
- Spojení:
- iFluor® 710
- iFluor® 710
- Klon:
- SK7
- SK7
- Reaktivita:
- Human
- Hostitel:
- Mouse
- Mouse
- ImunoFluorescence:
- Yes
- Yes
- Průtoková cytometrie:
- Yes
- Yes
- Čištění:
- Affinity chromatography
- Affinity chromatography
- Křížová adsorbce:
- No
- No
- Skladovací teplota:
- 2...8 °C with minimised light exposure, do not freeze
- 2...8 °C with minimised light exposure, do not freeze
- Transportní teplota:
- 20
- 20
Frequently Bought Together