Assistent Brush with Natural Bristles
Catalog # HECH91120000000
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Bristle material:Přírodní štětiny
- Kartáčová hlava Ø:13 mm
- Handle material:Ocelový drát
- Total length:900 mm
- Environmentally Preferable:
- Bal.:1
Specifications
About this item
Brush with natural bristles for use with laboratory glassware.
- Galvanised steel wire handle
- Natural bristles