Assistent Brush with Wooden Handle
Katalogové číslo HECH44279010
Dodavatel: GLASWARENFABRIK KARL HECHT
Specifikace
- Bristle material:Nylon
- Kartáčová hlava Ø:30 - 130 mm
- Handle material:Dřevo
- Total length:700 mm
- Environmentally Preferable:
- Bal.:1
Specifikace
O tomto produktu
Brushes for sedimentation vessels.
Cleaning brushes with wooden handles, designed for use with sedimentation vessels.