Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD44 Rabbit Monoclonal Antibody [clone: CD44v4/1700R]
Anti-CD44 Rabbit Monoclonal Antibody [clone: CD44v4/1700R]
Catalog # PRSI34-065
Supplier:  ProSci Inc.
Anti-CD44 Rabbit Monoclonal Antibody [clone: CD44v4/1700R]
Catalog # PRSI34-065
Supplier:  ProSci Inc.

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    CD44 molecule (Indian blood group)
  • Symbol antigenu:
    CD44
  • Klonovalita:
    Monoclonal
  • Klon:
    CD44v4/1700R
  • Spojení:
    Unconjugated
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG kappa
  • Formát:
    Purified Antibody
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    P16070
  • Synonyma antigenu:
    CD44
  • Skladovací pufr:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Skladovací teplota:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Koncentrace:
    0,2 mg/ml
  • Transportní teplota:
    4 °C
  • Imunogen:
    An amino acid sequence from the variant 4 domain of CD44 was used as the immunogen for the recombinant CD44v4 antibody.
  • Čištění:
    Protein A affinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifications

About this item

This antibody recognises an epitope encoded by exon v4 on the variant portion of human CD44. The CD44 molecule belongs to a family of cellular adhesion molecules found on a wide range of normal and malignant cells in epithelial, mesothelial and hemopoiesis tissues. It is a single gene with 20 exons, of which 10 are normally expressed to encode the basic CD44 (H-CAM) molecule. The additional 10 exons (v1 to v10) are only expressed by alternative splicing of the nuclear RNA. The expression of specific cell adhesion molecule CD44 splice variants has been reported to be associated with metastasis in certain human malignancies.

Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT Prediluted IHC only format: incubate for 30 min at RT (1) Titering of the recombinant CD44v4 antibody may be required for optimal performance.

1. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: CD44v4/1700R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG, kappa
Reactivity: Human