Zadání objednávky
Czech Republic
ContactUsLinkComponent
 
Anti-CD44 Mouse Monoclonal Antibody [clone: CD44v9/1459]
  PRSI33-652
New Product
Anti-CD44 Mouse Monoclonal Antibody [clone: CD44v9/1459]
  PRSI33-652
New Product

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    CD44 molecule (Indian blood group)
  • Symbol antigenu:
    CD44
  • Klonovalita:
    Monoclonal
  • Klon:
    CD44v9/1459
  • Spojení:
    Unconjugated
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG1 kappa
  • Formát:
    Purified Antibody
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    P16070
  • Synonyma antigenu:
    CD44
  • Skladovací pufr:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Skladovací teplota:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Koncentrace:
    0,2 mg/ml
  • Transportní teplota:
    4 °C
  • Imunogen:
    An amino acid sequence from the variant 9 domain of CD44 was used as the immunogen for the CD44v9 antibody.
  • Čištění:
    Protein G affinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

 

 

This antibody recognises an epitope encoded by exon v9 on the variant portion of human CD44. The CD44 molecule belongs to a family of cellular adhesion molecules found on a wide range of normal and malignant cells in epithelial, mesothelial and hemopoiesis tissues. CD44 is a single gene with 20 exons, of which 10 are normally expressed to encode the basic CD44 (H-CAM) molecule. The additional 10 exons (v1 to v10) are only expressed by alternative splicing of the nuclear RNA. The expression of specific cell adhesion molecule CD44 splice variants has been reported to be associated with metastasis in certain human malignancies.

Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT Titering of the CD44v9 antibody may be required for optimal performance.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: CD44v9/1459
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human