Specifications
- Druh protilátky:Primární
- Název antigenu:cyclin-dependent kinase inhibitor 1C (p57, Kip2)
- Symbol antigenu:CDKN1C
- Klonovalita:Monoclonal
- Klon:KIP2/880
- Spojení:Unconjugated
- Průtoková cytometrie:Yes
- Hostitel:Mouse
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Izotyp:IgG2b kappa
- Formát:Purified Antibody
- Formulář:Liquid
- Identifikační číslo genu:P49918
- Synonyma antigenu:CDKN1C
- Skladovací pufr:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Skladovací teplota:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Koncentrace:0,2 mg/ml
- Transportní teplota:4 °C
- Imunogen:Recombinant human protein was used as the immunogen for the p57 antibody.
- Čištění:Protein G affinity chromatography
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Recognises a protein of 57 kDa, identified as p57Kip2. It shows no cross-reaction with p27Kip1. p57Kip2 is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. Anti-p57 has been used as an aide in identification of complete hydatidiform mole (CHM) (no nuclear labeling of cytotrophoblasts and stromal cells) from partial hydatidiform mole (PHM) in which both cytotrophoblasts and stromal cells stain. The histological differentiation of complete mole, partial mole, and hydropic spontaneous abortion is problematic. Most complete hydatidiform moles are diploid, whereas most partial moles are triploid. Ploidy studies will identify partial moles, but will not differentiate complete moles from non-molar gestations. Complete moles carry a high risk of persistent disease and choriocarcinoma, while partial moles have a very low risk. In normal placenta, many cytotrophoblast nuclei and stromal cells are labeled with this antibody. Similar findings apply to PHM and hydropic abortus tissues. Intervillous trophoblastic islands (IVTIs) demonstrate nuclear labeling in all three entities and serve as an internal control.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,25-0,5 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Optimal dilution of the p57 antibody to be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CDKN1C
Clonality: Monoclonal
Clone: KIP2/880
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human, Mouse