Human Recombinant HABP1 (from E. coli)
Catalog # PRSI92-123
Supplier: ProSci Inc.
New Product
CAS Number:
Specifications
- Bal.:0,05 mg
- Typ proteinu/peptidu:Recombinant
- Zdroj:E. coli
- Druhy:Human
- Sekvence značek:C-6 His tag
- Podmínky uskladnění:Store at −20 °C, stable for 6 months after receipt. Please minimise freeze-thaw cycles.
- Obsah endotoxinů:<0,1 ng/µg (1 IEU/µg) as determined by LAL test
- Proteinová synonyma:GC1QBP|HABP1|SF2P32|C1qBP|Mitochondrial Matrix Protein p32|ASF/SF2-Associated Protein p32|gC1q-R Protein|p33|Hyaluronan-Binding Protein 1|Complement Component 1 Q Subcomponent-Binding Protein Mitochondrial|Glycoprotein gC1qBP
- Název proteinu/peptidu:HABP1
- Čistota:>95% as determined by reducing SDS-PAGE
- Molekulární hmotnost:24,9 kDa
- Sekvence:Leu74-Gln282
- Formulace:Supplied as a 0,2 µm filtered solution of 20 mM Tris, 20% Glycerol, 1 mM DTT, pH 7,5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the Lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
- Testované aplikace:Bioassay
Specifications
About this item
Complement Component 1Q Subcomponent-Binding Protein (C1QBP) is a nucleus protein that belongs to the MAM33 family. C1QBP is known to bind to the globular heads of C1q molecules and inhibit C1 activation. Mitochondrial C1QBP is a critical mediator of p14ARF-induced apoptosis. C1QBP functions as a chemotactic factor for immature dendritic cells, and migration is mediated through ligation of both C1QBP and cC1qR/CR. C1QBP overexpression successfully blocks mRNA accumulation from the adenovirus major late transcription unit (MLTU) and stimulates RNA polymerase II carboxy-terminal domain phosphorylation in virus-infected cells.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays.
Caution: For research use only.