Anti-MK-3475 Recombinant Antibody
Catalog # PRSI10-614
Supplier: ProSci Inc.
New Product
Specifications
- Druh protilátky:Primární
- Klonovalita:Recombinant
- Spojení:Unconjugated
- Izotyp:IgG1 lambda
- Synonyma antigenu:unconjugated:BAY 86-1903|BAY 94-9343
- Modifikace:None
- Skladovací pufr:PBS buffer pH 7,5
- Skladovací teplota:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
- Koncentrace:Batch dependent
- Transportní teplota:20
- Imunogen:Homo sapiens/MSLN [Homo sapiens]
- Čištění:>95%
- Typ balení:Vial
- Bal.:0,1 mg
Specifications
About this item
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-lambda
Reactivity: