Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CNTO 1275 Recombinant Antibody
Anti-CNTO 1275 Recombinant Antibody
Catalog # PRSI10-192
Supplier:  ProSci Inc.
Anti-CNTO 1275 Recombinant Antibody
Catalog # PRSI10-192
Supplier:  ProSci Inc.

Specifications

  • Druh protilátky:
    Primární
  • Klonovalita:
    Recombinant
  • Spojení:
    Unconjugated
  • Izotyp:
    IgG4 lambda
  • Synonyma antigenu:
    CAT-354
  • Modifikace:
    None
  • Skladovací pufr:
    PBS buffer pH 7,5
  • Skladovací teplota:
    Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
  • Koncentrace:
    Batch dependent
  • Transportní teplota:
    20
  • Imunogen:
    Homo sapiens/IL13 [Homo sapiens]
  • Čištění:
    >95%
  • Typ balení:
    Vial
  • Bal.:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4-lambda
Reactivity: