Order Entry
Czech Republic
ContactUsLinkComponent
Anti-EPCAM Recombinant Antibody [Clone: KS1/4]
Anti-EPCAM Recombinant Antibody [Clone: KS1/4]
Catalog # PRSI24-818
Supplier:  ProSci Inc.
New Product
Anti-EPCAM Recombinant Antibody [Clone: KS1/4]
Catalog # PRSI24-818
Supplier:  ProSci Inc.
New Product

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    epithelial cell adhesion molecule
  • Symbol antigenu:
    EPCAM
  • Klonovalita:
    Recombinant
  • Klon:
    KS1/4
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Průtoková cytometrie:
    Yes
  • Izotyp:
    IgG2a kappa
  • Reaktivita:
    Human
  • Křížová adsorbce:
    No
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    4072
  • Synonyma antigenu:
    CD326|Epithelial glycoprotein|Epithelial glycoprotein 314|Epithelial cell surface antigen|Ep-CAM|hEGP314|EGP314|KSA|KS1/4 antigen|Adenocarcinoma-associated antigen|Tumor-associated calcium signal transducer 1.|17-1A-antigen|EGP|Epithelial cell adhesion molecule|Cell surface glycoprotein Trop-1|Major gastrointestinal tumor-associated protein GA733-2
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    PBS with 0,02% Proclin 300
  • Skladovací teplota:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Transportní teplota:
    Shipped on blue ice
  • Imunogen:
    The original antibody was generated by immunizing BALB/c mice with human adenocarcinoma of the lung cell line UCLA P3
  • Testované aplikace:
    Immunoperoxidase staining
  • Typ balení:
    Vial
  • Bal.:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: epithelial cell adhesion molecule
Clonality: Recombinant
Clone: KS1/4
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Human