Plastic Box with White Lid
Catalog # MULC192-3000
Supplier: MULTIROIR
Specifications
- Type:Safety lock
- Kapacita:3 L
- Materiál:PP
- Š×H×V:193×193×112 mm
- Bal.:72
Specifications
About this item
These combi box containers are versatile, providing ideal storage solutions for a diverse range of laboratory applications.
- Polypropylene material
- Watertight lid
- Tamper-proof lid
- Stackable box
- Food contact quality
Ideal for laboratory environments, this versatile container ensures secure storage and transport of samples and reagents. The airtight, tamper-evident lid provides enhanced safety, while the stackable design maximises space efficiency. The opaque white polypropylene construction ensures durability and easy identification of contents.