Methylová oranž 0,1% in aqueous solution Reag. Ph. Eur. indicator, Fluka™
34576-500ML
:
- Typ balení:Plastová láhev
- Bal.:500 mL
- Sekvence:2725
- Test of sensitivity:complying
- Change range (pH 3,0-4,4):complying
:
:0,970 g/cm3 (20 °C)
:39 °C
:Ambient
:208-925-3
:1170
:3,III
:GHS02
:547-58-0
:C₁₄H₁₄N₃NaO₃S
:MFCD00007502