Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-cis-Diltiazem HCl
L-cis-Diltiazem HCl ≥98%
L-cis-Diltiazem HCl ≥98%
L-cis-Diltiazem HCl ≥98%
ENZOBMLCA2070005
:
Enzo Life Sciences
:
L-cis-Diltiazem HCl ≥98%
ENZOBMLCA2070005
:
Enzo Life Sciences
:
Bal.:
5 mg
Sekvence:
2210
Purity:
≥98%
:
:
451.0 g/mol
:
GHS07
:
Ambient
:
42399-54-2
:
C₂₂H₂₆N₂O₄S·HCl