ODN 2118 (Control for ODN 1585) (synthetic), TLRgrade™
Catalog # ENZOALX746203C100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Bal.:100 µG
- Sekvence:1286
Specifications
Specification Test Results
- Source:Synthetic
- Formulation:Lyophilised. Sterile.
- Purity:Activity and endotoxin tested - TLRgrade™
- Endotoxin content:<0.002 EU/µg
Specification Test Results
About this item
:
:6.643 g/mol
Storage Temperature:Refrigerator