Order Entry
Czech Republic
ContactUsLinkComponent
ODN 1585 (synthetic), TLRgrade™
ODN 1585 (synthetic), TLRgrade™
Catalog # ENZOALX746053M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
ODN 1585 (synthetic), TLRgrade™
Catalog # ENZOALX746053M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Bal.:
    1 mg
  • Sekvence:
    2070

Specifications

Specification Test Results

  • Source:
    Synthetic
  • Formulation:
    Lyophilised. Sterile.
  • Purity:
    Activity and endotoxin tested - TLRgrade™
  • Endotoxin content:
    <0.002 EU/µg
  • Quantity:
    150 nmol (~1000 µg)

Specification Test Results

About this item

 :
 :6.643 g/mol
Storage Temperature:Refrigerator