ContactUsLinkComponent
8-Isopentenylnaringenin ≥98% (by 1H-NMR)
8-Isopentenylnaringenin ≥98% (by 1H-NMR)
Catalog # ENZOALX385025M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
8-Isopentenylnaringenin ≥98% (by 1H-NMR)
Catalog # ENZOALX385025M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Bal.:
    5 mg
  • Sekvence:
    2210
Specifications
About this item
 :
 :340.4 g/mol
Storage Temperature:Refrigerator
CAS Number:68682-02-0
Formula:C₂₀H₂₀O₅