Order Entry
Czech Republic
ContactUsLinkComponent
Morin ≥85%
Morin ≥85%
Catalog # ENZOALX385016G001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Morin ≥85%
Catalog # ENZOALX385016G001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Bal.:
    1 g
  • Sekvence:
    3047

Specifications

About this item

 :
 :302.24 g/mol
CAS Number:480-16-0
 :MFCD00217054
Formula:C₁₅H₁₀O₇
MDL Number:MFCD00006826