UCN-01 (from Streptomyces sp. MST-AS5345) ≥99% (by HPLC)
Catalog # ENZOALX380222MC25
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Bal.:0,25 mg
- Sekvence:2025
Specifications
About this item
:
:482.5 g/mol
Storage Temperature:Freezer
CAS Number:112953-11-4
Formula:C₂₈H₂₆N₄O₄