Fostriecin sodium salt (from Streptomyces pulveraceous subsp. fostreus) ≥98% (by HPLC)
Katalogové číslo ENZOALX380065C010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Bal.:10 µG
- Sekvence:1106
Specifications
About this item
:
:452.37 g/mol
Storage Temperature:Freezer
CAS Number:87860-39-7
Formula:C₁₉H₂₆O₉PNa