0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Phomopsin A
Phomopsin A ≥98% (by HPLC)
Phomopsin A ≥98% (by HPLC)
Phomopsin A ≥98% (by HPLC)
Catalog # ENZOALX350417M001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Phomopsin A ≥98% (by HPLC)
Catalog # ENZOALX350417M001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Bal.:
1 mg
Sekvence:
2070
Specifications
About this item
Product Details
:
:
789.2 g/mol
CAS Number:
64925-80-0
Formula:
C₃₆H₄₅ClN₆O₁₂
Product Details