Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Oxamflatin
Oxamflatin ≥95% (by HPLC)
Oxamflatin ≥95% (by HPLC)
Oxamflatin ≥95% (by HPLC)
Catalog # ENZOALX270379M005
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Oxamflatin ≥95% (by HPLC)
Catalog # ENZOALX270379M005
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Bal.:
5 mg
Sekvence:
2210
Specifications
About this item
Product Details
:
CAS Number:
151720-43-3
Product Details