EQ0 NHS ester amine-reactive quencher dye
Katalogové číslo ENZOENZCHM2080001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Bal.:1 mg
- Sekvence:2070
Specifications
Specification Test Results
- Excitation maximum:445 nm
- Emission maximum:none
- Extinction Coefficient:23,600 M-1cm-1
- Correction Factor (260 nm):0.48
- Correction Factor (280 nm):0.42
- Purity:≥80% (HPLC)
- Appearance:Red brown solid
Specification Test Results
About this item
:
:439.5 g/mol