Order Entry
Czech Republic
ContactUsLinkComponent
Ibotenic acid ≥98% (by HPLC), white to off-white solid
Toxic
Ibotenic acid ≥98% (by HPLC), white to off-white solid
Catalog # ENZOBMLEA1200001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Toxic
Ibotenic acid ≥98% (by HPLC), white to off-white solid
Catalog # ENZOBMLEA1200001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Bal.:
    1 mg
  • Sekvence:
    2070

Specifications

Specification Test Results

  • Identity:
    Determined by H-NMR and MS
  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :158.113 g/mol
Storage Temperature:Ambient
UN:3462
ADR:6.1,III
 :GHS06
CAS Number:2552-55-8
Formula:C₅H₆N₂O₄
MDL Number:MFCD00069294