Order Entry
Czech Republic
ContactUsLinkComponent
12(R)-Hydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HETE) ≥99%
12(R)-Hydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HETE) ≥99%
Catalog # ENZOBMLH1120050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
12(R)-Hydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HETE) ≥99%
Catalog # ENZOBMLH1120050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Bal.:
    50 µG
  • Sekvence:
    1202

Specifications

About this item

 :
 :320.5 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
CAS Number:82337-46-0
Formula:C₂₀H₃₂O₃