ContactUsLinkComponent
Damnacanthal ≥95% (by HPLC)
Damnacanthal ≥95% (by HPLC)
  ENZOBMLEI2740001
 :  
undefined
Damnacanthal ≥95% (by HPLC)
  ENZOBMLEI2740001
 :  
  • Bal.:
    1 mg
  • Sekvence:
    2070
 :
 :282.2 g/mol
 :Freezer
 :477-84-9
 :C₁₆H₁₀O₅