ContactUsLinkComponent
Leukotriene B4-dimethylamide ≥98% (by HPLC)
Leukotriene B4-dimethylamide ≥98% (by HPLC)
Katalogové číslo ENZOBMLEI1020050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Leukotriene B4-dimethylamide ≥98% (by HPLC)
Katalogové číslo ENZOBMLEI1020050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Bal.:
    50 µG
  • Sekvence:
    1202
Specifications
About this item
 :
 :363.5 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
CAS Number:83024-92-4
Formula:C₂₂H₃₇NO₃