0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Domů
S-Methyl-L-thiocitrulline dihydrochloride
S-Methyl-L-thiocitrulline dihydrochloride ≥97%
S-Methyl-L-thiocitrulline dihydrochloride ≥97%
S-Methyl-L-thiocitrulline dihydrochloride ≥97%
Catalog # ENZOALX106012M010
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
S-Methyl-L-thiocitrulline dihydrochloride ≥97%
Catalog # ENZOALX106012M010
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Bal.:
10 mg
Sekvence:
2275
Specifications
About this item
Product Details
:
CAS Number:
209589-59-3
Product Details