ContactUsLinkComponent
Ilimaquinone ≥98% (by TLC)
Ilimaquinone ≥98% (by TLC)
Catalog # ENZOBMLG4350100
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Ilimaquinone ≥98% (by TLC)
Catalog # ENZOBMLG4350100
Supplier:  Enzo Life Sciences
CAS Number:  
Specifications
  • Bal.:
    100 µG
  • Sekvence:
    1286
Specifications
About this item
 :
 :358.478 g/mol
CAS Number:71678-03-0
Formula:C₂₂H₃₀O₄
MDL Number:MFCD22200891