0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Meloxicam
Meloxicam ≥98%
Meloxicam ≥98%
Catalog # ENZOBMLEI2920100
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Rozbalit obrázek
Meloxicam ≥98%
Catalog # ENZOBMLEI2920100
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Bal.:
100 mg
Sekvence:
2440
Specifications
About this item
Product Details
:
:
351.40 g/mol
CAS Number:
71125-38-7
Formula:
C₁₄H₁₃N₃O₄S₂
MDL Number:
MFCD00868752
Product Details