Neg-50™ Frozen Section Medium
Neg-50™ Medium, Modrá, 118 ml, 2
Katalogové číslo MENZ6502B
Dodavatel: Epredia
Přejít na:
Product Details & Documents
Yield high-quality sections down to –50 °C without chatter using Epredia™ Neg-50™ frozen section medium, a water-soluble frozen section medium.
- Buffered to ensure proper alkalinity (pH = 8,0)
- Specially formulated to prevent tissue separation from the slide
This colourless, water-soluble frozen section medium yields high-quality sections down to –50 °C without chatter. It leaves no residue after staining, and is compatible with the Microm® Automatic Cryo Approach system.
Specifikace
Specifikace
- Bal.:2
- Barva:Modrá
- Popis:Neg-50™ Medium
- Objem:118 ml