NANOCOLOR® ECO Photometric Reagent Kit, Iron
Catalog # MANA976002
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Bal.:1 KIT
- Popis:Iron
- Rozměry:148×148×115 mm
- Metoda:Iron - 1, 10 - phenanthroline
- Plošina:Nanocolour eco reagent kits
- Skladovací teplota:15 - 25 °C
- Parametry:Iron
- Stupnice měření:Iron - 0,01 - 5,00 mg/l
- Životnost police (měsíce):18 months
- Hmotnost:425,4 g
- Testů v kitu:100
Specifications
About this item
Reagent kit for the determination of iron. Precise rapid tests for all kind of water and waste water samples. Time saving and reliable analysis together with our NANOCOLOR photometers.
- Sea water analysis
- Shelf life for 1,5 year(s)
- Reagents for 100 tests in a box
Iron is ubiquitous in our daily lives. Iron salts are essential for human life and are absorbed through food and drinking water. However, excessive concentrations of iron in water are undesirable due to discoloration and the formation of deposits such as rust.