Mycoplasma removal agent, MP Biomedicals
ICNA093050044
:
- Bal.:5 mL
- Popis:Mycoplasma removal agent
Mycoplasma removal agent (MRA) is an effective solution for the problem of mycoplasma contamination in cell cultures.
- Completely kills mycoplasma with no recurrence of contamination
- Effective at low concentration
MRA is a derivative of the quinoline family of antibiotics. It eliminates mycoplasma infection (recommended use at a concentration of 0,5 μg/ml) by inhibiting mycoplasma DNA gyrase. Cell toxicity is low following treatment at the recommended concentration, as is regrowth of mycoplasma after cellular transfer.
: Supplied as a ready to use solution and each vial can decontaminate up to 25 cell cultures.