xPIPETTE™ Carousel Stand
Catalog # 613-6912
Supplier: BIOTIX
Specifications
- For:For xPIPETTE™ pipettes
- Popis:Carousel stand
- Materiál:PP
- Environmentally Preferable:
- Bal.:1
Specifications
About this item
The carousel pipette stand is a rotating carousel stand for the xPIPETTE.
- Fits both single and multichannel models
- Holds seven pipettes total
- Made from chemically resistant polypropylene
Pipettes hang vertically, preventing liquids from contaminating the pipette shaft.