ECHO Cellcyte 3 Live Cell Imaging and Analysis
Live cell imaging and analysis in real-time from within the incubator.
- Enhanced phase contrast: High resolution and contrast rich transmission imaging mode
- Fluorescence microscopy: Three channel (DAPI, GFP, mCherry) or four channel (DAPI, GFP, RFP, CY5) available
- Open fanless design: easier accessibility and maintenance; uses convection to heat and circulate warm air, eliminating vibration and the formation of hot pockets
- Acquisition software: Automated acquisition and user friendly image and data visualisation software
- Analysis software: Integrated real-time image analysis using smart algorithms
- Analysis modules: Confluence, cell counting, apoptosis, cytotoxicity, transfection efficiency, reporter genes, immune cell cluster formations
The Cellcyte 3 is suitable for a variety of cell culture vessels and can support up to six wellplates (6 to 384 wells each) and T-series flasks. 8 TB internal storage. Additional storage can be added via external hard drives or through locally attached network drives. Ultra-bright LEDs with 50000 - hour lifespan.
5 MP Monochrome camera: increases field of view and resolution for fluorescence imaging
Operating conditions: (Imaging unit) 10 to 40 °C, RH up to 95%, (Controller unit) 20 to 40 °C
Illumination technique: Enhanced contour and fluorescence imaging (three or four fluorescence channels available)
Export formats: Movie (AVI), image (TIFF, PNG), data (CVS, XLXS)
: To be completed with control unit and flask inserts, which are sold separately. Control unit with software (required option) - for device control and acquisition. Spheroid software (optional) - real time Spheroid imaging and analysis at 4X.
: Supplied with Cellcyte 3 imaging unit, control unit, integrated monochrome camera for enhanced phase contrast and fluorescence imaging, and power supply.
Live cell imaging and analysis in real-time from within the incubator.
- Enhanced phase contrast: High resolution and contrast rich transmission imaging mode
- Fluorescence microscopy: Three channel (DAPI, GFP, mCherry) or four channel (DAPI, GFP, RFP, CY5) available
- Open fanless design: easier accessibility and maintenance; uses convection to heat and circulate warm air, eliminating vibration and the formation of hot pockets
- Acquisition software: Automated acquisition and user friendly image and data visualisation software
- Analysis software: Integrated real-time image analysis using smart algorithms
- Analysis modules: Confluence, cell counting, apoptosis, cytotoxicity, transfection efficiency, reporter genes, immune cell cluster formations
The Cellcyte 3 is suitable for a variety of cell culture vessels and can support up to six wellplates (6 to 384 wells each) and T-series flasks. 8 TB internal storage. Additional storage can be added via external hard drives or through locally attached network drives. Ultra-bright LEDs with 50000 - hour lifespan.
5 MP Monochrome camera: increases field of view and resolution for fluorescence imaging
Operating conditions: (Imaging unit) 10 to 40 °C, RH up to 95%, (Controller unit) 20 to 40 °C
Illumination technique: Enhanced contour and fluorescence imaging (three or four fluorescence channels available)
Export formats: Movie (AVI), image (TIFF, PNG), data (CVS, XLXS)
: To be completed with control unit and flask inserts, which are sold separately. Control unit with software (required option) - for device control and acquisition. Spheroid software (optional) - real time Spheroid imaging and analysis at 4X.
: Supplied with Cellcyte 3 imaging unit, control unit, integrated monochrome camera for enhanced phase contrast and fluorescence imaging, and power supply.
- Catalog No:
- ECHLECCC3T25
- ECHLECCC3T75
- ECHLECCC3CU3
- ECHLECCC3233
- Kontrastní technika:
- -
- Fluorescence and phase contrast
- Zvětšení:
- —
- 4×, 10×, 20×
- Kamera:
- —
- 5 MP sCMOS monochrome camera (2448×2048 pixels)
- Snímač:
- —
- CMOS
- Displej:
- —
- 55,88 cm (22") HD monitor
- Výstup:
- —
- USB
- Elektrické napětí:
- —
- 100 - 240 V (50/60 Hz)
Product Family Options
Flask inserts
- SekvencePopisOsvětlení
Specifications
Cat. No.ECHLECCC3T25Kontrastní technikaZvětšeníKameraSnímačDisplejVýstupElektrické napětíSpecifications
Cat. No.ECHLECCC3T75Kontrastní technikaZvětšeníKameraSnímačDisplejVýstupElektrické napětí
- SekvencePopisOsvětlení
- 03Cellcyte 3 Control unit (PC and 55,88 cm (22") HD monitor) with software for device control and acquisition1 * 1 KS572 000,00 Kč
Specifications
Cat. No.ECHLECCC3CU3Kontrastní technika-Zvětšení—Kamera—Snímač—Displej—Výstup—Elektrické napětí— - 01Cellcyte 3 Inverted imaging and analysis system, three-channel fluorescenceDAPI - EX: 395/25 EM:450/30;
GFP - EX:483/18 EM:525/25;
mCherry - EX:585/22 EM:625/40 (fluorescence options)1 * 1 KS1 987 700,00 KčNew ProductSpecifications
Cat. No.ECHLECCC3233Kontrastní technikaFluorescence and phase contrastZvětšení4×, 10×, 20×Kamera5 MP sCMOS monochrome camera (2448×2048 pixels)SnímačCMOSDisplej55,88 cm (22") HD monitorVýstupUSBElektrické napětí100 - 240 V (50/60 Hz)New Product
Objective lenses
- SekvenceZvětšeníPopisPracovní vzdálenost
- 2320×Fluorite ELWD objective6,6 - 7,8 mm1 * 1 KS57 030,00 Kč