Supelco® BIOshell™ 400 Å Protein C18 Fused-Core® HPLC Columns
Supplier: Merck
Certificates
About this item
BIOshell™ 400 Å Protein C18 columns contain 3,4 µm advanced Fused-Core® particles with 400 Å pores that are specifically engineered to provide efficient separation of large proteins.
- A 0,2 µm-thick pourous shell with 400 Å pores leads to rapid and efficient separation of proteins
- Superficial porous particles (SPPs) provide narrower peak widths and imprved resolution for characterisation of biomolecules in comparison to fully porous particles (FPPs)
- Ability to tolerate high temperature applications (up to 90 °C) in acidic mobile phase
- Compatible with UHPLC, HPLC and MS
These columns offer maximum hydrophobicity for resolving tricky hydrophilic and PEGylated proteins.
Certifications: Column test certificate available inside the box.
Accessories information: This guard cartridge holder can be used with 4,6/3,0 and 2,1 mm Int.Ø BIOShell™ columns.
Specifications
- Catalog No:
- 67474-U
- 67506-U
- 67475-U
- 67472-U
- 67505-U
- 67473-U
- 67477-U
- 67457-U
- 67458-U
- 67478-U
- 67496-U
- 67482-U
- 67459-U
- 67508-U
- 67487-U
- 67469-U
- 67488-U
- 67471-U
- 67483-U
- 67462-U
- 67485-U
- 67463-U
- Aplikace:
- mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments - mAbs
ADC's
Biosimilars/Proteins/mAb Fragments
- Naplnění uhlíkem:
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- 1,00%
- S navazanou skupinou:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Maximální tlak:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- Rozsah pH:
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- Povrchová oblast:
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- 15 m²/g
- Teplotní limit:
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- Kód USP:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1