ContactUsLinkComponent
Accessories for Dissolved Oxygen Meter, Bench, inoLab® Oxi 7310
Accessories for Dissolved Oxygen Meter, Bench, inoLab® Oxi 7310
Katalogové číslo WTWA205210
Dodavatel:  WTW
Accessories for Dissolved Oxygen Meter, Bench, inoLab® Oxi 7310
Katalogové číslo WTWA205210
Dodavatel:  WTW
Specifikace
  • Popis:
    Electrolyte for amperometric oxygen sensors, 1 bottle with 50 ml
  • Environmentally Preferable:
  • Bal.:
    50 mL