Electronic Overhead Stirrers, Hei-TORQUE Silver Package
Catalog # HEID501-61019-00
Supplier: Heidolph Instruments GmbH & Co.KG
Specifications
- Popis:Hei-TORQUE Silver package, EU plug
- Sekvence:02
- Bal.:1
Specifications
About this item
Hei-TORQUE Silver package with the perfect end-to-end solution for powerful stirring and the simplest operation in the laboratory.
- Hei-TORQUE Expert 100
- Telescope stand
- Clamp
Packaging: Each package contains a telescopic stand and a suitable clamp for easy use on lab tables.