Manual Vacuum Regulation Turret, Flush Mount
Katalogové číslo ILMV700562
Dodavatel: Welch by Gardner Denver
Specifikace
- Popis:Manual vacuum regulation valve, flush mount, 2 screw mount, for use with wood furniture
- Bal.:1
Specifikace
O tomto produktu
Open/close the vacuum line and easily control flow rate.
- Dimensions: 690 mm
- Diameter: 820 mm protrusion from wall and 20 mm into wall
Turret has stepped hose barb that accepts 6,35 - 9,52 mm vacuum hose (DN 8-10).