CryoPlus™ Rack for Cryogenic Vials
Catalog # THBR4000005
Supplier: Thermo Fisher Scientific
Specifications
- Určeno pro:CryoPlus 1, 2, 3, 4
- Popis:Square rack, vapor phase
- Bal.:1
Specifications
About this item
CryoPlus™ series racks are ideal for use with CryoPlus™ LN2 storage systems.
- Rack includes: cardboard boxes and cardboard dividers for 5 ml vials
- 6 cardboard boxes with 100 cell divider
- Holds: 6 boxes/rack