Gel Load Pipette Tips
1 - 200 µl, Přírodní, Packed in a resealable bag
Katalogové číslo GREI775390_10000
Dodavatel: Greiner Bio-One
Přejít na:
Product Details & Documents
Gel-load pipette tips with a slim shape that allows even the thinnest gels to be loaded without damaging them. Due to the long and flexible tip, pipetting at the bottom of very narrow vessels is also possible without any problems.
- Made of polypropylene
- Universal fit for all common pipettes
- 5 µl graduation for easy control of the pipetted volume
- Tip with 0,57 mm outer diameter suitable for loading gel pockets
- 83 mm total length to reach even the bottom of narrow vessels
The gel-load pipette tips are packed in a resealable bag, which means that the tips are safely protected from possible contamination even after the bag has been opened.
Free of detectable DNase, RNase, human DNA, pyrogens and PCR inhibitors.
Specifikace
- Barevný kód:Přírodní
- Balené:Packed in a resealable bag
- Objem:1 - 200 µl