Orders LinkContactUsLinkComponent
Mobile Tool Cabinets
Mobile Tool Cabinets
Dodavatel:   TEKNOMEK
Mobile Tool Cabinets
Dodavatel:  TEKNOMEK
Certifikáty
Mobile Tool Cabinets
2 Dostupných variant níže
Product Details & Documents
This heavy duty mobile stainless steel tool cabinet with 3, 4 or 11 drawers has a brushed dull polish finish is excellent for tool storage within the workplace. With a floor clearance of 100 mm, there is plenty of room for a mop or broom for easy cleaning below the tool cabinet.
  • Easy tool organisation
  • Fully welded 304L-grade stainless steel
  • Brushed dull polish finish
  • Stainless steel castors (2 fixed and 2 locking swivel)
  • Lockable drawers
Designed with lockable drawers of varying sizes this storage cabinet provides easy and safe compartmentalisation of different types of tools for efficiency and safety.
Product Family Options
Informace o produktu
  • Sekvence
    Materiál
    Š×H×V
    Dostupnost
    Price
  • TEKMSHCCCATO4D1050
    Mobile tool cabinet with 4 drawers
    Mobile tool cabinet with 4 drawers
    01
    Stainless steel 304L
    1000×500×840 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS
  • TEKMSHCCCATO111050
    Mobile tool cabinet with 11 drawers
    Mobile tool cabinet with 11 drawers
    02
    Stainless steel 304L
    1050×500×1040 mm