Glass Stopper, Octagonal
24/29, Brown glass
Katalogové číslo NEUB1641-92-2429
Dodavatel: NEUBERT VOLUME GLASSWAERE
Přejít na:
Product Details & Documents
Solid glass stopper made of borosilicate glass 3.3 octagonal.
- Eight sided
- Diameter: 60 mm
Specifikace
- NZ:24/29
- Type:Brown glass
- Cat. No.:NEUB1641-92-2429