Hydrogenfosforečnan didraselný 99.0-101.0% (by titrimetric analysis), LiChropur™ suitable for HPLC, Supelco®
Catalog # 59678-250G
Supplier: Merck
CAS Number:
Specifications
- Bal.:250 g
- Sekvence:3545
Specifications
About this item
:
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383